The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023292.4 | NP_075781.3 | 3 - 4 | 1033 | 151 |
GenBank mRNA | AF266505.1 | - | 2 - 3 | 987 | 151 |
AK030639.1 | - | 3 - 4 | 1429 | 151 | |
AK079095.1 | - | 3 - 4 | 1034 | 151 | |
BC029253.1 | - | 2 - 3 | 994 | 151 |