Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Srpr
Entrez Gene ID
67398
Gene Name
signal recognition particle receptor ('docking protein')
Gene Aliases
1300011P19Rik, D11Mgi27
Chromosome Location
Chr.9: 35211203 - 35217003 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC012512.1 - - 1167 150

View Full Product Details