Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Srpr
Entrez Gene ID
67398
Gene Name
signal recognition particle receptor ('docking protein')
Gene Aliases
1300011P19Rik, D11Mgi27
Chromosome Location
Chr.9: 35211203 - 35217003 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026130.1 NP_080406.1 7 - 8 1015 81
GenBank mRNA AK004967.2 - 7 - 8 1015 81
AK038017.1 - 7 - 8 1003 81
AK206633.1 - 2 - 3 197 81
BC012512.1 - - 98 81
BC021839.1 - 7 - 8 998 81

View Full Product Details