Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Snx22
Entrez Gene ID
382083
Gene Name
sorting nexin 22
Gene Aliases
Gm1120
Chromosome Location
Chr.9: 66065176 - 66069780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025612.2 NP_001020783.1 6 - 7 510 63
GenBank mRNA AK032028.1 - 6 - 7 499 63
BC138950.1 - 6 - 7 497 63
BC138951.1 - 6 - 7 497 63

View Full Product Details