Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rdh8
Entrez Gene ID
235033
Gene Name
retinol dehydrogenase 8
Gene Aliases
Gm182, prRDH
Chromosome Location
Chr.9: 20818504 - 20826163 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030290.1 NP_001025461.1 2 - 3 264 61

View Full Product Details