Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mns1
Entrez Gene ID
17427
Gene Name
meiosis-specific nuclear structural protein 1
Gene Aliases
AW546487
Chromosome Location
Chr.9: 72438518 - 72458576 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008613.3 NP_032639.1 9 - 10 1548 79
GenBank mRNA AK013048.1 - 9 - 10 1501 79
AK132772.1 - 9 - 10 1523 79
BC046624.1 - 9 - 10 1461 79
D14849.1 - 9 - 10 1579 79

View Full Product Details