Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oaf
Entrez Gene ID
102644
Gene Name
OAF homolog (Drosophila)
Gene Aliases
AL024221, D130038B21Rik, D9Ucla1
Chromosome Location
Chr.9: 43221278 - 43239816 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178644.3 NP_848759.1 2 - 3 638 93
GenBank mRNA AK051349.1 - 2 - 3 638 93
AK150279.1 - 2 - 3 625 93
AK193808.1 - 1 - 2 126 93
BC025514.1 - 2 - 3 646 93
BC026022.1 - 2 - 3 602 93
BC028824.1 - 2 - 3 673 93
BC034836.1 - 2 - 3 604 93

View Full Product Details