Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scg3
Entrez Gene ID
20255
Gene Name
secretogranin III
Gene Aliases
1B1075, AI385542, Chgd, SgIII
Chromosome Location
Chr.9: 75643366 - 75684082 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164790.1 NP_001158262.1 8 - 9 1322 141
NM_009130.3 NP_033156.1 8 - 9 1322 141
GenBank mRNA AK133655.1 - 8 - 9 1224 141
AK139140.1 - 8 - 9 1262 141
AK158315.1 - 8 - 9 1195 141
AK164089.1 - 8 - 9 1265 141
BC024785.1 - 8 - 9 1281 141
U02982.1 - 8 - 9 1286 141

View Full Product Details