The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025857.2 | NP_080133.1 | 9 - 10 | 919 | 130 |
GenBank mRNA | AK009203.1 | - | 3 - 4 | 253 | 130 |
AK017714.1 | - | 9 - 10 | 885 | 130 | |
AK159897.1 | - | 9 - 10 | 920 | 130 | |
BC027311.1 | - | 9 - 10 | 915 | 130 |