The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028854.3 | NP_083130.1 | 9 - 10 | 943 | 57 |
GenBank mRNA | AK007086.1 | - | 9 - 10 | 1031 | 57 |
BC100468.1 | - | 9 - 10 | 1009 | 57 | |
BC132329.1 | - | 9 - 10 | 943 | 57 | |
BC138332.1 | - | 9 - 10 | 943 | 57 | |
BC145239.1 | - | 9 - 10 | 940 | 57 | |
BC145240.1 | - | 9 - 10 | 992 | 57 |