Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stoml1
Entrez Gene ID
69106
Gene Name
stomatin-like 1
Gene Aliases
1810015E19Rik, AI847431, SLP1, STORP, UNC24, WPB72
Chromosome Location
Chr.9: 58253164 - 58262524 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026942.3 NP_081218.3 2 - 3 339 77
GenBank mRNA AK045820.1 - 2 - 3 311 77
AK148324.1 - - 238 77
BC037074.1 - 2 - 3 269 77

View Full Product Details