Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Nxpe4
- Entrez Gene ID
-
244853
- Gene Name
- neurexophilin and PC-esterase domain family, member 4
- Gene Aliases
-
C130036J11, D930028F11Rik, Fam55d
- Chromosome Location
-
Chr.9: 48162001 - 48400025 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK048132.1 |
- |
2 - 3 |
313 |
72 |
View Full Product Details