The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172921.3 | NP_766509.2 | 7 - 8 | 1464 | 117 |
GenBank mRNA | AK048132.1 | - | 8 - 9 | 1476 | 117 |
AK136478.1 | - | 3 - 4 | 858 | 117 | |
BC085195.1 | - | 3 - 4 | 1287 | 117 | |
BC094249.1 | - | 9 - 10 | 1663 | 117 |