Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Isl2
Entrez Gene ID
104360
Gene Name
insulin related protein 2 (islet 2)
Gene Aliases
3110001N10Rik
Chromosome Location
Chr.9: 55538677 - 55547667 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027397.3 NP_081673.2 4 - 5 931 57
GenBank mRNA BC120880.2 - 3 - 4 611 57

View Full Product Details