The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_180600.3 | NP_850931.2 | 1 - 2 | 457 | 143 |
GenBank mRNA | AK042515.1 | - | 1 - 2 | 458 | 143 |
AK083216.1 | - | 1 - 2 | 450 | 143 | |
AK150776.1 | - | 1 - 2 | 282 | 143 | |
BC029111.1 | - | 1 - 2 | 286 | 143 |