The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027830.2 | NP_082106.2 | 13 - 14 | 2424 | 68 |
GenBank mRNA | AK133212.1 | - | 6 - 7 | 726 | 68 |
AK133345.1 | - | 6 - 7 | 1539 | 68 | |
BC062185.1 | - | 4 - 5 | 501 | 68 |