The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170800.1 | NP_001164271.1 | 8 - 9 | 1250 | 82 |
GenBank mRNA | AK038257.1 | - | 2 - 3 | 345 | 82 |
AK220339.1 | - | 7 - 8 | 1071 | 82 |