Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eml6
Entrez Gene ID
237711
Gene Name
echinoderm microtubule associated protein like 6
Gene Aliases
2900083P10Rik, C230094A16Rik, EMAP-6
Chromosome Location
Chr.11: 29742586 - 30026034 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146016.2 NP_666128.2 33 - 34 5361 72
GenBank mRNA BC024726.1 - 5 - 6 796 72

View Full Product Details