The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159603.1 | NP_001153075.1 | 14 - 15 | 2439 | 64 |
NM_001159604.1 | NP_001153076.1 | 14 - 15 | 2439 | 64 | |
NM_001159605.1 | NP_001153077.1 | 12 - 13 | 2151 | 64 | |
NM_001159606.1 | NP_001153078.1 | 10 - 11 | 1713 | 64 | |
NM_030722.2 | NP_109647.2 | 14 - 15 | 2442 | 64 | |
GenBank mRNA | AF321909.1 | - | 14 - 15 | 2412 | 64 |
AK122205.1 | - | 14 - 15 | 2422 | 64 | |
AK141921.1 | - | 14 - 15 | 2439 | 64 | |
AK147185.1 | - | 10 - 11 | 1685 | 64 | |
AK154059.1 | - | 13 - 14 | 2037 | 64 | |
AK161135.1 | - | 12 - 13 | 2145 | 64 | |
AK163977.1 | - | 14 - 15 | 2430 | 64 | |
BC048174.1 | - | 14 - 15 | 2392 | 64 | |
BC050747.1 | - | 14 - 15 | 2412 | 64 |