Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pum1
Entrez Gene ID
80912
Gene Name
pumilio RNA-binding family member 1
Gene Aliases
AA517475, Pumm, mKIAA0099
Chromosome Location
Chr.4: 130663297 - 130781564 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159603.1 NP_001153075.1 20 - 21 3354 76
NM_001159604.1 NP_001153076.1 20 - 21 3360 76
NM_001159605.1 NP_001153077.1 18 - 19 3072 76
NM_001159606.1 NP_001153078.1 16 - 17 2628 76
NM_030722.2 NP_109647.2 20 - 21 3363 76
GenBank mRNA AF321909.1 - 20 - 21 3333 76
AK122205.1 - 20 - 21 3343 76
AK141921.1 - 20 - 21 3360 76
AK147185.1 - 16 - 17 2600 76
AK154059.1 - 19 - 20 2952 76
AK161135.1 - 18 - 19 3066 76
AK162742.1 - 19 - 20 3050 76
AK163977.1 - 20 - 21 3351 76
BC048174.1 - 20 - 21 3313 76
BC050747.1 - 20 - 21 3327 76

View Full Product Details