Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ulbp1
Entrez Gene ID
77777
Gene Name
UL16 binding protein 1
Gene Aliases
A430108B07Rik, MULT1
Chromosome Location
Chr.10: 7444873 - 7473637 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029975.2 NP_084251.1 5 - 6 994 89
GenBank mRNA AK020784.1 - 5 - 6 869 89
AK084712.1 - - 1093 89
AK085119.1 - - 1156 89
AY249021.1 - 4 - 5 623 89
BC104292.1 - 2 - 3 241 89
BC104293.1 - 2 - 3 241 89
BC114390.1 - 2 - 3 240 89
BC114420.1 - 2 - 3 240 89
BC127022.1 - 2 - 3 241 89
BC127023.1 - 2 - 3 241 89
BC132070.1 - 5 - 6 994 89
BC132072.1 - 5 - 6 994 89

View Full Product Details