The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146155.3 | NP_666267.3 | 2 - 3 | 113 | 101 |
GenBank mRNA | AK149347.1 | - | 2 - 3 | 360 | 101 |
BC042621.1 | - | 2 - 3 | 113 | 101 | |
BC060231.1 | - | 1 - 2 | 81 | 101 |