Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam13a
Entrez Gene ID
58909
Gene Name
family with sequence similarity 13, member A
Gene Aliases
AI462012, D430015B01Rik, FAM13A1, Precm1
Chromosome Location
Chr.6: 58933536 - 59024502 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153574.2 NP_705802.1 17 - 18 2123 155
GenBank mRNA AJ510136.1 - 17 - 18 2109 155
AK036291.1 - 10 - 11 1330 155
AK052421.1 - 17 - 18 2064 155
AK087913.1 - - 337 155
BC080861.1 - 17 - 18 2123 155

View Full Product Details