Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam13a
Entrez Gene ID
58909
Gene Name
family with sequence similarity 13, member A
Gene Aliases
AI462012, D430015B01Rik, FAM13A1, Precm1
Chromosome Location
Chr.6: 58933536 - 59024502 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153574.2 NP_705802.1 9 - 10 1114 95
GenBank mRNA AJ510136.1 - 9 - 10 1100 95
AK036291.1 - 2 - 3 321 95
AK052421.1 - 9 - 10 1055 95
BC005554.1 - 9 - 10 1019 95
BC054053.1 - 9 - 10 1088 95
BC060978.1 - 9 - 10 1051 95
BC080861.1 - 9 - 10 1114 95

View Full Product Details