The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253392.1 | NP_001240321.1 | 18 - 19 | 2480 | 76 |
NM_007958.1 | NP_031984.1 | 20 - 21 | 2941 | 76 | |
GenBank mRNA | AK122454.1 | - | 20 - 21 | 2748 | 76 |
BC042442.1 | - | 18 - 19 | 2473 | 76 | |
CF531941.1 | - | 2 - 3 | 224 | 76 |