Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrpl19
Entrez Gene ID
56284
Gene Name
mitochondrial ribosomal protein L19
Gene Aliases
9030416F12Rik, 9130412E02Rik, D6Ertd157e, MRP-L15, RLX1, Rpml15
Chromosome Location
Chr.6: 81957826 - 81965949 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026490.2 NP_080766.1 2 - 3 356 61
GenBank mRNA AK021180.1 - 2 - 3 350 61
AK030793.1 - 2 - 3 344 61
AK050637.1 - 2 - 3 355 61
AK141554.1 - 2 - 3 366 61
BC020315.2 - 2 - 3 332 61
BC043921.1 - 2 - 3 337 61
BC061064.1 - 2 - 3 340 61

View Full Product Details