Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Txnrd3
Entrez Gene ID
232223
Gene Name
thioredoxin reductase 3
Gene Aliases
AI196535, TR2, Tgr
Chromosome Location
Chr.6: 89643988 - 89675529 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178058.1 NP_001171529.1 15 - 16 2034 81
NM_001178059.1 NP_001171530.1 13 - 14 1692 81
NM_001178060.1 NP_001171531.1 13 - 14 1692 81
NM_153162.3 NP_694802.2 15 - 16 2034 81
GenBank mRNA AF349659.1 - 15 - 16 1943 81
AK012699.1 - 15 - 16 2027 81
AK041046.1 - 15 - 16 2037 81
AK080362.1 - 15 - 16 2025 81
AK134987.1 - 13 - 14 1683 81
AK207943.1 - 1 - 2 135 81
BC076605.1 - 15 - 16 1992 81

View Full Product Details