The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178058.1 | NP_001171529.1 | 6 - 7 | 885 | 134 |
NM_001178059.1 | NP_001171530.1 | 6 - 7 | 885 | 134 | |
NM_001178060.1 | NP_001171531.1 | 6 - 7 | 885 | 134 | |
NM_153162.3 | NP_694802.2 | 6 - 7 | 885 | 134 | |
GenBank mRNA | AF349659.1 | - | 6 - 7 | 794 | 134 |
AK012699.1 | - | 6 - 7 | 878 | 134 | |
AK041046.1 | - | 6 - 7 | 887 | 134 | |
AK080362.1 | - | 6 - 7 | 876 | 134 | |
AK134987.1 | - | 6 - 7 | 876 | 134 | |
AK190297.1 | - | 1 - 2 | 99 | 134 | |
BC076605.1 | - | 6 - 7 | 843 | 134 |