Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chchd6
Entrez Gene ID
66098
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 6
Gene Aliases
0710001P09Rik, 1700021B03Rik
Chromosome Location
Chr.6: 89383146 - 89595652 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167736.1 NP_001161208.1 6 - 7 846 86
NM_025351.3 NP_079627.3 7 - 8 930 86
GenBank mRNA AK002980.1 - 7 - 8 914 86
AK005696.1 - 7 - 8 862 86
AK006192.1 - 7 - 8 866 86
AK015314.1 - 7 - 8 893 86
AK087642.1 - 7 - 8 840 86
AK144093.1 - 7 - 8 836 86
BC011331.1 - 7 - 8 893 86
BC052929.1 - 7 - 8 889 86
EF410100.1 - - 990 86

View Full Product Details