Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtf1
Entrez Gene ID
17764
Gene Name
metal response element binding transcription factor 1
Gene Aliases
MTF-1, Thyls
Chromosome Location
Chr.4: 124802549 - 124849800 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008636.4 NP_032662.3 10 - 11 1976 62
GenBank mRNA AK030284.1 - 10 - 11 1955 62
AK150295.1 - 11 - 12 2033 62
AY260849.1 - 9 - 10 1827 62
AY260851.1 - 2 - 3 185 62
AY260852.1 - 2 - 3 193 62
BC017679.1 - 10 - 11 1938 62

View Full Product Details