Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mtf1
Entrez Gene ID
17764
Gene Name
metal response element binding transcription factor 1
Gene Aliases
MTF-1, Thyls
Chromosome Location
Chr.4: 124802549 - 124849800 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008636.4 NP_032662.3 8 - 9 1314 76
GenBank mRNA AK030284.1 - 8 - 9 1293 76
AK150295.1 - 9 - 10 1374 76
AY260849.1 - 7 - 8 1168 76
BC017679.1 - 8 - 9 1276 76
X71327.1 - 8 - 9 1264 76

View Full Product Details