Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crbn
Entrez Gene ID
58799
Gene Name
cereblon
Gene Aliases
2610203G15Rik, 2900045O07Rik, AF229032, AW108261, piL
Chromosome Location
Chr.6: 106778244 - 106800087 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021449.3 NP_067424.2 5 - 6 724 110
NM_175357.3 NP_780566.1 5 - 6 727 110
GenBank mRNA AF229032.1 - 5 - 6 679 110
AK032836.1 - 5 - 6 701 110
AK050557.1 - 5 - 6 710 110
AK075959.1 - 5 - 6 701 110
AK076144.1 - 5 - 6 707 110
AK077707.1 - 5 - 6 707 110
AK160219.1 - 5 - 6 703 110
BC033458.1 - 5 - 6 685 110
BC046967.1 - 4 - 5 642 110
BC069905.1 - 5 - 6 684 110
BC086488.1 - 5 - 6 702 110

View Full Product Details