Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Uba3
Entrez Gene ID
22200
Gene Name
ubiquitin-like modifier activating enzyme 3
Gene Aliases
A830034N06Rik, AI256736, AI848246, AW546539, Ube1c
Chromosome Location
Chr.6: 97183632 - 97205647 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF077330.1 - 13 - 14 1060 123
AK032514.1 - 14 - 15 1294 123
AK048148.1 - 14 - 15 1092 123
AK159381.1 - 14 - 15 1180 123
AK166655.1 - 13 - 14 1050 123
AK168859.1 - 14 - 15 1095 123
AY029181.1 - 13 - 14 1060 123
BC002002.1 - 14 - 15 1080 123
BC080776.1 - 13 - 14 1013 123
CF928750.1 - 3 - 4 193 123

View Full Product Details