Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aldh1l1
Entrez Gene ID
107747
Gene Name
aldehyde dehydrogenase 1 family, member L1
Gene Aliases
1810048F20Rik, FDH, Fthfd, Neut2
Chromosome Location
Chr.6: 90550767 - 90599171 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027406.1 NP_081682.1 17 - 18 2131 62
GenBank mRNA BC022112.1 - 16 - 17 2082 62
BC024055.1 - 17 - 18 2088 62
BC025939.1 - 17 - 18 2113 62
BC028817.1 - 17 - 18 2078 62
BC030722.1 - 17 - 18 2115 62
BC030723.1 - 17 - 18 2047 62
BC030727.1 - 17 - 18 2117 62
BC030730.1 - 17 - 18 2131 62

View Full Product Details