The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167730.1 | NP_001161202.1 | 3 - 4 | 266 | 157 |
NM_021385.2 | NP_067360.2 | 3 - 4 | 266 | 157 | |
GenBank mRNA | AF205278.1 | - | 3 - 4 | 190 | 157 |
AK012795.1 | - | 3 - 4 | 267 | 157 | |
AK028505.1 | - | 3 - 4 | 186 | 157 | |
AK030043.1 | - | 3 - 4 | 267 | 157 | |
AK048975.1 | - | 3 - 4 | 241 | 157 | |
BC011120.1 | - | 3 - 4 | 229 | 157 |