Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oxtr
Entrez Gene ID
18430
Gene Name
oxytocin receptor
Gene Aliases
OTR
Chromosome Location
Chr.6: 112473684 - 112491308 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081147.1 NP_001074616.1 1 - 2 934 93
GenBank mRNA AK143340.1 - 1 - 2 899 93
D86599.1 - 3 - 4 1487 93

View Full Product Details