Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ggcx
Entrez Gene ID
56316
Gene Name
gamma-glutamyl carboxylase
Gene Aliases
-
Chromosome Location
Chr.6: 72414296 - 72431106 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019802.5 NP_062776.1 14 - 15 2182 72
GenBank mRNA AF087938.1 - 14 - 15 2157 72
AK033496.1 - 14 - 15 2109 72
AK036577.1 - 14 - 15 2156 72
AK135425.1 - 14 - 15 2169 72
AK165982.1 - 14 - 15 2157 72
BC138763.1 - 14 - 15 2107 72
BC138764.1 - 14 - 15 2107 72

View Full Product Details