The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144917.5 | NP_659166.1 | 2 - 3 | 124 | 85 |
GenBank mRNA | AK049155.1 | - | 2 - 3 | 54 | 85 |
BY074266.1 | - | 3 - 4 | 147 | 85 |