Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sh2d6
Entrez Gene ID
71130
Gene Name
SH2 domain containing 6
Gene Aliases
-
Chromosome Location
Chr.6: 72513652 - 72534207 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK016886.1 - 11 - 12 859 96
BC100525.1 - 11 - 12 816 96

View Full Product Details