Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptcd3
Entrez Gene ID
69956
Gene Name
pentatricopeptide repeat domain 3
Gene Aliases
2610034F17Rik, 2810422B04Rik, AA589622, AU045708, MRP-S39
Chromosome Location
Chr.6: 71880638 - 71908772 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027275.3 NP_081551.2 19 - 20 1561 96
GenBank mRNA AK013131.1 - 5 - 6 340 96
AK146005.1 - 19 - 20 1550 96
AK146172.1 - 19 - 20 1562 96
AK172480.1 - 20 - 21 1626 96
BC026759.1 - 3 - 4 216 96
BC055844.1 - 5 - 6 388 96
BC060687.1 - 10 - 11 782 96
BC084595.1 - 10 - 11 811 96
BC115434.2 - 19 - 20 1557 96

View Full Product Details