Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ntf3
Entrez Gene ID
18205
Gene Name
neurotrophin 3
Gene Aliases
AI316846, AI835689, Nt3, Ntf-3
Chromosome Location
Chr.6: 126101412 - 126166791 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164034.1 NP_001157506.1 2 - 3 318 98
NM_001164035.1 NP_001157507.1 1 - 2 233 98
GenBank mRNA AA198264.1 - 2 - 3 318 98
AK132586.1 - 1 - 2 233 98
BC065785.1 - 1 - 2 194 98

View Full Product Details