The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198604.2 | NP_941006.2 | 14 - 15 | 1968 | 60 |
GenBank mRNA | AK153619.1 | - | 14 - 15 | 2258 | 60 |
BC026778.1 | - | 14 - 15 | 1971 | 60 |