The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033122.3 | NP_001028294.1 | 4 - 5 | 580 | 101 |
GenBank mRNA | AK151076.1 | - | 4 - 5 | 581 | 101 |
AK153224.1 | - | 4 - 5 | 581 | 101 | |
BC106997.2 | - | 4 - 5 | 565 | 101 | |
BC128272.1 | - | 4 - 5 | 565 | 101 | |
BC128273.1 | - | 4 - 5 | 565 | 101 | |
BC139135.1 | - | 4 - 5 | 542 | 101 | |
BC139136.1 | - | 4 - 5 | 542 | 101 |