Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klra7
Entrez Gene ID
16638
Gene Name
killer cell lectin-like receptor, subfamily A, member 7
Gene Aliases
LGL-1, Ly-49G.1, Ly-49G.2, Ly-49G.3, Ly49G.4, Ly49g, Ly49g1, Ly49g2
Chromosome Location
Chr.6: 130218605 - 130233322 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF074457.1 - 2 - 3 208 90
AF283248.1 - 2 - 3 208 90
AF283249.1 - 2 - 3 208 90
AF288375.1 - 2 - 3 201 90
AF288376.1 - 2 - 3 230 90
AF307945.1 - 2 - 3 208 90
AF307946.1 - 2 - 3 208 90
AF419249.1 - - 490 90
BC099859.1 - 2 - 3 207 90
BC099860.1 - 2 - 3 207 90
BC099862.1 - 2 - 3 207 90
BC130251.1 - 2 - 3 256 90
BC140984.1 - 2 - 3 254 90
GU434662.1 - 2 - 3 201 90
GU434663.1 - 2 - 3 201 90
S78689.1 - 2 - 3 230 90

View Full Product Details