The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205364.1 | NP_001192293.1 | 1 - 2 | 187 | 61 |
GenBank mRNA | EU339279.1 | - | 1 - 2 | 94 | 61 |