The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178703.4 | NP_848818.1 | 13 - 14 | 1810 | 75 |
GenBank mRNA | AK042956.1 | - | 13 - 14 | 1672 | 75 |
AK052971.1 | - | 13 - 14 | 1635 | 75 | |
AK053883.1 | - | 13 - 14 | 1632 | 75 | |
BC059080.1 | - | 13 - 14 | 1661 | 75 | |
M92378.1 | - | 12 - 13 | 1569 | 75 |