The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026894.1 | NP_081170.1 | 6 - 7 | 971 | 64 |
GenBank mRNA | AK005100.1 | - | 6 - 7 | 971 | 64 |
AK165459.1 | - | 2 - 3 | 462 | 64 | |
BE956902.1 | - | 2 - 3 | 64 |