Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aak1
Entrez Gene ID
269774
Gene Name
AP2 associated kinase 1
Gene Aliases
5530400K14Rik, 9630042K20, AU067724, AU067726, BC028270, C79663, D6Ertd245e, R75501, mKIAA1048
Chromosome Location
Chr.6: 86849482 - 87003227 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040106.2 NP_001035195.1 9 - 10 1588 73
NM_177762.6 NP_808430.2 9 - 10 1588 73
GenBank mRNA AK147363.1 - 9 - 10 1589 73
BC141176.1 - 9 - 10 1344 73
CF723295.1 - 2 - 3 108 73

View Full Product Details