The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040106.2 | NP_001035195.1 | 1 - 2 | 694 | 115 |
NM_177762.6 | NP_808430.2 | 1 - 2 | 694 | 115 | |
GenBank mRNA | AK147363.1 | - | 1 - 2 | 695 | 115 |
AK158879.1 | - | 1 - 2 | 584 | 115 | |
BC141176.1 | - | 1 - 2 | 450 | 115 |