Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Aak1
Entrez Gene ID
269774
Gene Name
AP2 associated kinase 1
Gene Aliases
5530400K14Rik, 9630042K20, AU067724, AU067726, BC028270, C79663, D6Ertd245e, R75501, mKIAA1048
Chromosome Location
Chr.6: 86849482 - 87003227 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040106.2 NP_001035195.1 3 - 4 933 70
NM_177762.6 NP_808430.2 3 - 4 933 70
GenBank mRNA AK147363.1 - 3 - 4 934 70
AK158879.1 - 3 - 4 823 70
BC141176.1 - 3 - 4 689 70
CA528349.1 - 1 - 2 102 70

View Full Product Details